siRNA Details
| Gene Symbol: Olr1730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000279.1 | NP_001000279.1 | 1 | 94 | ||
| Gene Symbol: Olr1869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213732.1 | NP_998897.1 | 1 | 94 | ||

Olr1730
Genomic Map