siRNA Details
| Gene Symbol: Olr1481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000527.1 | NP_001000527.1 | 1 | 314 | ||
| Gene Symbol: LOC103693363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767910.1 | XP_008766132.1 | 1 | 314 | ||

Olr1481
Genomic Map