siRNA Details
| Gene Symbol: Olr916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000587.1 | NP_001000587.1 | 1 | 598 | ||
| Gene Symbol: Olr1845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240816.1 | XP_006240878.1 | 1 | 598 | ||

Olr916
Genomic Map