siRNA Details
| Gene Symbol: Olr786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000916.1 | NP_001000916.1 | 1 | 151 | ||
| Gene Symbol: LOC100909613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749539.1 | XP_003749587.1 | 151 | |||

Olr786
Genomic Map