siRNA Details
| Gene Symbol: Olr1686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001373.1 | NP_001001373.1 | 1 | 910 | ||
| Gene Symbol: LOC100910479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255857.2 | XP_006255919.1 | 1243 | |||

Olr1686
Genomic Map