siRNA Details
| Gene Symbol: Ugt2b15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004271.1 | NP_001004271.1 | 6 | 1679 | ||
| GenBank mRNA | BC078782.1 | - | 6 | 1679 | 7107952 | |
| FQ219334.1 | - | 6 | 1173 | |||
| U06273.1 | - | 6 | 1660 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2b15| Gene Symbol: Ugt2b15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004271.1 | NP_001004271.1 | 6 | 1679 | ||
| GenBank mRNA | BC078782.1 | - | 6 | 1679 | 7107952 | |
| FQ219334.1 | - | 6 | 1173 | |||
| U06273.1 | - | 6 | 1660 | |||