siRNA Details
| Gene Symbol: Rassf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007754.1 | NP_001007755.1 | 2 | 274 | ||
| NM_001037555.1 | NP_001032644.1 | 3 | 466 | |||
| GenBank mRNA | AB202124.1 | - | 3 | 466 | ||
| BC079237.1 | - | 2 | 274 | 7132629 | ||
| FQ233012.1 | - | 2 | 314 | |||

Rassf1
Genomic Map