siRNA Details
| Gene Symbol: Esrrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008516.2 | NP_001008516.2 | 4, 5 | 785 | ||
| XM_006240374.2 | XP_006240436.1 | 5, 6 | 1247 | |||
| XM_006240375.2 | XP_006240437.1 | 5, 6 | 1248 | |||
| XM_008764759.1 | XP_008762981.1 | 5, 6 | 2483 | |||
| GenBank mRNA | AY383731.1 | - | 4, 5 | 785 | ||

Esrrb
Genomic Map