siRNA Details
| Gene Symbol: V1ra14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008898.1 | NP_001008898.1 | 1 | 193 | ||
| GenBank mRNA | AY510301.1 | - | 1 | 193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

V1ra14| Gene Symbol: V1ra14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008898.1 | NP_001008898.1 | 1 | 193 | ||
| GenBank mRNA | AY510301.1 | - | 1 | 193 | ||