siRNA Details
| Gene Symbol: Vom1r93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008971.1 | NP_001008971.1 | 1 | 722 | ||
| XM_006236848.1 | XP_006236910.1 | 3 | 1058 | |||
| GenBank mRNA | AY510293.1 | - | 1 | 722 | ||
| U36896.1 | - | 3 | 1072 | |||

Vom1r93
Genomic Map