siRNA Details
| Gene Symbol: Ly49s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009488.1 | NP_001009488.1 | 3, 4 | 319 | ||
| GenBank mRNA | AY649837.1 | - | 3, 4 | 319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49s6| Gene Symbol: Ly49s6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009488.1 | NP_001009488.1 | 3, 4 | 319 | ||
| GenBank mRNA | AY649837.1 | - | 3, 4 | 319 | ||