siRNA Details
| Gene Symbol: RAB22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020673.2 | NP_065724.1 | 7 | 804 | ||
| GenBank mRNA | AF091034.2 | - | 7 | 782 | ||
| AK091180.1 | - | 7 | 558 | |||
| BC007036.1 | - | 3 | 191 | |||
| BC015710.1 | - | 7 | 729 | 3907891 | ||
| BC063457.1 | - | 7 | 788 | 5182786 | ||
| BT007046.1 | - | 7 | 542 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024436, AJ311124, AK150617, BC006596, AK149200, and others.

RAB22A
Genomic Map