siRNA Details
| Gene Symbol: Ly49i4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009495.1 | NP_001009495.1 | 2 | 240 | ||
| XM_008763405.1 | XP_008761627.1 | 3 | 355 | |||
| GenBank mRNA | AY649834.1 | - | 2 | 240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49i4| Gene Symbol: Ly49i4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009495.1 | NP_001009495.1 | 2 | 240 | ||
| XM_008763405.1 | XP_008761627.1 | 3 | 355 | |||
| GenBank mRNA | AY649834.1 | - | 2 | 240 | ||