siRNA Details
| Gene Symbol: Ly49i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009499.1 | NP_001009499.1 | 85 | |||
| GenBank mRNA | AY659932.1 | - | 85 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49i3| Gene Symbol: Ly49i3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009499.1 | NP_001009499.1 | 85 | |||
| GenBank mRNA | AY659932.1 | - | 85 | |||