siRNA Details
| Gene Symbol: Sub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009618.1 | NP_001009618.1 | 2 | 146 | ||
| XM_006232043.2 | XP_006232105.1 | 2 | 113 | |||
| GenBank mRNA | BC088346.1 | - | 2 | 146 | 7368325 | |
| FQ211681.1 | - | 2 | 103 | |||
| FQ211748.1 | - | 2 | 103 | |||
| FQ230867.1 | - | 2 | 76 | |||
| FQ232387.1 | - | 2 | 75 | |||

Sub1
Genomic Map