siRNA Details
| Gene Symbol: Oas1i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009680.1 | NP_001009680.1 | 4, 5 | 915 | ||
| GenBank mRNA | AY196697.1 | - | 4, 5 | 915 | ||
| BC160819.1 | - | 4, 5 | 889 | 7382468 | ||
| Gene Symbol: LOC100910735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751197.2 | XP_003751245.1 | 935 | |||

Oas1i
Genomic Map