siRNA Details
| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009692.1 | NP_001009692.1 | 4 | 423 | ||
| GenBank mRNA | BC087652.1 | - | 4 | 423 | 7312960 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sh3glb2| Gene Symbol: Sh3glb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009692.1 | NP_001009692.1 | 4 | 423 | ||
| GenBank mRNA | BC087652.1 | - | 4 | 423 | 7312960 | |