siRNA Details
| Gene Symbol: Metrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009962.1 | NP_001009962.1 | 4 | 824 | ||
| GenBank mRNA | AY800384.1 | - | 4 | 824 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Metrn| Gene Symbol: Metrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009962.1 | NP_001009962.1 | 4 | 824 | ||
| GenBank mRNA | AY800384.1 | - | 4 | 824 | ||