siRNA Details
| Gene Symbol: Spetex-2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009969.1 | NP_001009969.1 | 4 | 511 | ||
| GenBank mRNA | AB180083.1 | - | 4 | 511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spetex-2H| Gene Symbol: Spetex-2H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009969.1 | NP_001009969.1 | 4 | 511 | ||
| GenBank mRNA | AB180083.1 | - | 4 | 511 | ||