siRNA Details
| Gene Symbol: LOC494539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY856054.1 | - | 2 | 407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC494539| Gene Symbol: LOC494539 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY856054.1 | - | 2 | 407 | ||