siRNA Details
| Gene Symbol: Fastk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006235892.2 | XP_006235954.1 | 3 | 763 | ||
| XM_006235894.2 | XP_006235956.1 | 3 | 750 | |||
| XM_006235895.1 | XP_006235957.1 | 3 | 747 | |||
| XM_006235896.2 | XP_006235958.1 | 3 | 764 | |||
| GenBank mRNA | BC083715.1 | - | 3 | 767 | 7190690 | |
| FQ215763.1 | - | 4 | 625 | |||
| FQ228972.1 | - | 3 | 686 | |||

Fastk
Genomic Map