siRNA Details
| Gene Symbol: Mad2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012106.1 | NP_001012106.1 | 9 | 838 | ||
| XM_006239390.2 | XP_006239452.1 | 11 | 1007 | |||
| XM_006239391.2 | XP_006239453.1 | 8 | 1377 | |||
| XM_008764256.1 | XP_008762478.1 | 10 | 1244 | |||
| GenBank mRNA | BC087687.1 | - | 9 | 838 | 7307981 | |

Mad2l2
Genomic Map