siRNA Details
| Gene Symbol: Ankrd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012112.1 | NP_001012112.1 | 3 | 1210 | ||
| XM_008764936.1 | XP_008763158.1 | 2 | 1758 | |||
| XM_008764937.1 | XP_008763159.1 | 3 | 1085 | |||
| GenBank mRNA | BC061827.1 | - | 1 | 993 | 5599008 | |
| BC079192.1 | - | 3 | 1210 | 7131342 | ||

Ankrd9
Genomic Map