siRNA Details
| Gene Symbol: Pcdhgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012215.1 | NP_001012215.1 | 1 | 619 | ||
| GenBank mRNA | BC087084.1 | - | 1 | 619 | 7189313 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhgb7| Gene Symbol: Pcdhgb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012215.1 | NP_001012215.1 | 1 | 619 | ||
| GenBank mRNA | BC087084.1 | - | 1 | 619 | 7189313 | |