siRNA Details
| Gene Symbol: Acsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325244.1 | - | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acsm5| Gene Symbol: Acsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325244.1 | - | 341 | |||