siRNA Details
| Gene Symbol: Lig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024268.2 | NP_001019439.1 | 26 | 2685 | ||
| GenBank mRNA | BC093604.1 | - | 26 | 2669 | 7317599 | |
| Gene Symbol: LOC100911727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759094.1 | XP_008757316.1 | 22 | 2354 | ||

Lig1
Genomic Map