siRNA Details
| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024270.2 | NP_001019441.2 | 17 | 3415 | ||
| GenBank mRNA | BC101890.1 | - | 17 | 3415 | 7930679 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pelp1| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024270.2 | NP_001019441.2 | 17 | 3415 | ||
| GenBank mRNA | BC101890.1 | - | 17 | 3415 | 7930679 | |