siRNA Details
| Gene Symbol: Psma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017280.2 | NP_058976.1 | 11 | 1660 | ||
| GenBank mRNA | BC081817.1 | - | 11 | 1660 | 7110009 | |
| FQ224319.1 | - | 1687 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psma3| Gene Symbol: Psma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017280.2 | NP_058976.1 | 11 | 1660 | ||
| GenBank mRNA | BC081817.1 | - | 11 | 1660 | 7110009 | |
| FQ224319.1 | - | 1687 | ||||