siRNA Details
| Gene Symbol: Ins2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019130.2 | NP_062003.1 | 2 | 167 | ||
| GenBank mRNA | BP472188.1 | - | 2 | 142 | ||
| BP485158.1 | - | 2 | 166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ins2| Gene Symbol: Ins2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019130.2 | NP_062003.1 | 2 | 167 | ||
| GenBank mRNA | BP472188.1 | - | 2 | 142 | ||
| BP485158.1 | - | 2 | 166 | |||