siRNA Details
| Gene Symbol: Tuba1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022298.1 | NP_071634.1 | 4 | 1445 | ||
| GenBank mRNA | BC062238.1 | - | 4 | 1479 | 6922126 | |
| BC078830.1 | - | 4 | 1458 | 7115494 | ||
| FQ210110.1 | - | 4 | 1477 | |||
| FQ212756.1 | - | 4 | 1412 | |||
| FQ213186.1 | - | 4 | 1478 | |||
| FQ214446.1 | - | 2 | 1034 | |||
| FQ216135.1 | - | 1 | 892 | |||
| FQ216180.1 | - | 1 | 892 | |||
| V01227.1 | - | 4 | 1445 | |||

Tuba1a
Genomic Map