siRNA Details
| Gene Symbol: Lig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF276774.1 | - | 26 | 2715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lig1| Gene Symbol: Lig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF276774.1 | - | 26 | 2715 | ||