siRNA Details
| Gene Symbol: Scgb2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080770.3 | 2 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb2a1| Gene Symbol: Scgb2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080770.3 | 2 | 268 | |||