siRNA Details
| Gene Symbol: Micalcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182669.2 | NP_872610.2 | 10 | 2098 | ||
| GenBank mRNA | AY149342.1 | - | 7 | 1538 | ||
| BC098647.1 | - | 10 | 2098 | 7112444 | ||
| BC100137.1 | - | 7 | 1552 | 7457561 | ||

Micalcl
Genomic Map