siRNA Details
| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183056.2 | NP_898879.2 | 11 | 1370 | ||
| XM_006238881.2 | XP_006238943.1 | 11 | 3080 | |||
| GenBank mRNA | BC100062.1 | - | 11 | 1370 | 7124923 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk40| Gene Symbol: Stk40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183056.2 | NP_898879.2 | 11 | 1370 | ||
| XM_006238881.2 | XP_006238943.1 | 11 | 3080 | |||
| GenBank mRNA | BC100062.1 | - | 11 | 1370 | 7124923 | |