siRNA Details
| Gene Symbol: Zfp85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001130.2 | NP_001001130.1 | 4 | 1913 | ||
| GenBank mRNA | BC055337.1 | - | 4 | 1913 | 6413269 | |
| BC055756.1 | - | 4 | 1913 | 6413269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp85| Gene Symbol: Zfp85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001130.2 | NP_001001130.1 | 4 | 1913 | ||
| GenBank mRNA | BC055337.1 | - | 4 | 1913 | 6413269 | |
| BC055756.1 | - | 4 | 1913 | 6413269 | ||