siRNA Details
| Gene Symbol: Wdr83os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001493.2 | NP_001001493.1 | 4 | 504 | ||
| GenBank mRNA | AK010893.1 | - | 4 | 506 | ||
| AK147129.1 | - | 4 | 728 | |||
| BC024937.1 | - | 4 | 501 | |||
| BC056474.1 | - | 4 | 474 | 6823629 | ||
| BC089623.1 | - | 4 | 504 | 30118707 | ||
| BC091742.1 | - | 4 | 447 | |||

Wdr83os
Genomic Map