siRNA Details
Gene Symbol: Zfp960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005358.2 | NP_001005358.2 | 4 | 2663 | ||
NM_001163919.1 | NP_001157391.1 | 5 | 1486 | |||
GenBank mRNA | AK145357.1 | - | 5 | 1474 | ||
BC018101.1 | - | 4 | 2653 | 3494875 | ||
BC064743.1 | - | 5 | 982 | |||
BC066177.1 | - | 5 | 982 |
Gene Symbol: Zfp97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011765.5 | NP_035895.3 | 4 | 2714 | ||
GenBank mRNA | DV660800.1 | - | 1 | 545 |
