siRNA Details
| Gene Symbol: Olfr111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005485.2 | NP_001005485.1 | 1 | 900 | ||
| GenBank mRNA | BC139116.1 | - | 1 | 900 | 8862138 | |
| BC139117.1 | - | 1 | 900 | 8862139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr111| Gene Symbol: Olfr111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005485.2 | NP_001005485.1 | 1 | 900 | ||
| GenBank mRNA | BC139116.1 | - | 1 | 900 | 8862138 | |
| BC139117.1 | - | 1 | 900 | 8862139 | ||