siRNA Details
| Gene Symbol: AY761184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 453 | ||
| GenBank mRNA | AK008498.1 | - | 2 | 459 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AY761184| Gene Symbol: AY761184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 453 | ||
| GenBank mRNA | AK008498.1 | - | 2 | 459 | ||