siRNA Details
| Gene Symbol: Olfr1251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011529.1 | NP_001011529.1 | 1 | 335 | ||
| GenBank mRNA | BC147159.1 | - | 1 | 391 | 9056474 | |
| BC147160.1 | - | 1 | 435 | 9056173 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1251| Gene Symbol: Olfr1251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011529.1 | NP_001011529.1 | 1 | 335 | ||
| GenBank mRNA | BC147159.1 | - | 1 | 391 | 9056474 | |
| BC147160.1 | - | 1 | 435 | 9056173 | ||