siRNA Details
Gene Symbol: Olfr723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011530.2 | NP_001011530.2 | 1 | 938 | ||
GenBank mRNA | BC150980.1 | - | 1 | 938 | 9087893 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011530.2 | NP_001011530.2 | 1 | 938 | ||
GenBank mRNA | BC150980.1 | - | 1 | 938 | 9087893 |