siRNA Details
| Gene Symbol: Olfr180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011662.2 | NP_001011662.2 | 2 | 947 | ||
| Gene Symbol: 4933431I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017005.1 | - | 3 | 974 | ||

Olfr180
Genomic Map