siRNA Details
| Gene Symbol: Rnase13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011687.2 | NP_001011687.1 | 1 | 239 | ||
| GenBank mRNA | BC132614.1 | - | 1 | 239 | 40130891 | |
| BC132616.1 | - | 1 | 239 | 40130893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnase13| Gene Symbol: Rnase13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011687.2 | NP_001011687.1 | 1 | 239 | ||
| GenBank mRNA | BC132614.1 | - | 1 | 239 | 40130891 | |
| BC132616.1 | - | 1 | 239 | 40130893 | ||