siRNA Details
| Gene Symbol: Olfr1066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011735.2 | NP_001011735.2 | 1 | 61 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1066| Gene Symbol: Olfr1066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011735.2 | NP_001011735.2 | 1 | 61 | ||