siRNA Details
| Gene Symbol: Olfr205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011736.1 | NP_001011736.1 | 1 | 14 | ||
| GenBank mRNA | BC150839.1 | - | 1 | 39 | 9087750 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr205| Gene Symbol: Olfr205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011736.1 | NP_001011736.1 | 1 | 14 | ||
| GenBank mRNA | BC150839.1 | - | 1 | 39 | 9087750 | |