siRNA Details
Gene Symbol: Olfr317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147131.1 | - | 1 | 1002 | 9056194 | |
BC147132.1 | - | 1 | 1002 | 9056174 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147131.1 | - | 1 | 1002 | 9056194 | |
BC147132.1 | - | 1 | 1002 | 9056174 |