siRNA Details
| Gene Symbol: Olfr222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011789.1 | NP_001011789.1 | 1 | 497 | ||
| GenBank mRNA | BC147220.1 | - | 1 | 572 | 9056253 | |
| BC147221.1 | - | 1 | 572 | 9056247 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr222| Gene Symbol: Olfr222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011789.1 | NP_001011789.1 | 1 | 497 | ||
| GenBank mRNA | BC147220.1 | - | 1 | 572 | 9056253 | |
| BC147221.1 | - | 1 | 572 | 9056247 | ||