siRNA Details
| Gene Symbol: Olfr1382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011790.1 | NP_001011790.1 | 1 | 308 | ||
| GenBank mRNA | BC119435.1 | - | 1 | 308 | 40041463 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1382| Gene Symbol: Olfr1382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011790.1 | NP_001011790.1 | 1 | 308 | ||
| GenBank mRNA | BC119435.1 | - | 1 | 308 | 40041463 | |