siRNA Details
| Gene Symbol: Olfr212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011800.2 | NP_001011800.1 | 2 | 1756 | ||
| GenBank mRNA | AK029829.1 | - | 2 | 1758 | ||
| BC141000.1 | - | 2 | 1565 | 9055612 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr212| Gene Symbol: Olfr212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011800.2 | NP_001011800.1 | 2 | 1756 | ||
| GenBank mRNA | AK029829.1 | - | 2 | 1758 | ||
| BC141000.1 | - | 2 | 1565 | 9055612 | ||