siRNA Details
| Gene Symbol: Olfr119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011830.2 | NP_001011830.2 | 2 | 594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr119| Gene Symbol: Olfr119 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011830.2 | NP_001011830.2 | 2 | 594 | ||